3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine sodium salt ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D4463-1G D4463-5G
TCD4463-1GEA 271.76 CAD
TCD4463-1G TCD4463-5G
3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine sodium salt ≥98.0% (by HPLC, total nitrogen)
3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine sodium salt
Formula: C₄₁H₄₅N₂NaO₆S₂
MDL Number: MFCD16621036
CAS Number: 64285-36-5

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Absorbance (E1%1cm) min. 2800 (MeOH, 675 to 685nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR