2,6-Dibromo-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D4384-1G
TCD4384-1GEA 279.34 CAD
TCD4384-1G
2,6-Dibromo-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)
2,6-Dibromo-1,4-phenylenediamine

CAS Number: 29213-03-4
Molecular Formula: C6H6Br2N2
Molecular Weight: 265.94
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138
Lambda max.: 334 nm (CH3CN)


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,5-Diamino-1,3-dibromobenzene

Formula: C₆H₆Br₂N₂
Melting Pt: 138 °C
CAS Number: 29213-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55002ee476ab8437a8a427af84502da691e0825d&vid=55002ee476ab8437a8a427af84502da691e0825d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR