Doxofylline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4302-1G D4302-25G
TCD4302-1GEA 58.46 CAD
TCD4302-1G TCD4302-25G
Doxofylline ≥98.0% (by HPLC, titration analysis)
Doxofylline

CAS Number: 69975-86-6
MDL Number: MFCD00865218
Molecular Formula: C11H14N4O4
Molecular Weight: 266.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients, Nucleobases and Their Analogs, Purine Alkaloids
Other Product Names / Synonyms: 7-(1,3-Dioxolan-2-ylmethyl)theophylline, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethyl-1H-purine-2,6-dione, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethylxanthine

Formula: C₁₁H₁₄N₄O₄
Melting Pt: 144 °C
MDL Number: MFCD00865218
CAS Number: 69975-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc8fe3abe989ec20fc5b727efd51de0e0c5ce71&vid=6dc8fe3abe989ec20fc5b727efd51de0e0c5ce71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR