4-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC)

Supplier: TCI America

D4291-1G D4291-5G
TCD4291-1GEA 198.29 CAD
TCD4291-1G TCD4291-5G
4-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC)
4-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene

CAS Number: 1173788-58-3
Molecular Formula: C22H39BO2S
Molecular Weight: 378.42
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-(4-Dodecyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Dodecyl-2-thiopheneboronic Acid Pinacol Ester

Formula: C₂₂H₃₉BO₂S
Density: 0.96 (20 °C)
CAS Number: 1173788-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb242af4a389d8329144d151c3ced541c0fb9735&vid=bb242af4a389d8329144d151c3ced541c0fb9735&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR