Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II) ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

D4191-1G
TCD4191-1GEA 339.7 CAD
TCD4191-1G
Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II) ≥97.0% (by HPLC, titration analysis)
Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II)

CAS Number: 847616-85-7
Molecular Formula: C24H20Cl2N2Pd2
Molecular Weight: 620.18
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 207

Formula: C₂₄H₁₈Cl₂N₂Pd₂
Melting Pt: 207 °C
MDL Number: MFCD22200542
CAS Number: 847616-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4b24a1eb595ecc48070682a8e7bce15cb6c8807&vid=e4b24a1eb595ecc48070682a8e7bce15cb6c8807&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR