9,10-Di(2-naphthyl)anthracene ≥98.0%

Supplier: TCI America

Synonyms: 9,10-Di(naphthalen-2-yl)anthracene

D4127-1G
TCD4127-1GEA 259.44 CAD
TCD4127-1G
9,10-Di(2-naphthyl)anthracene ≥98.0%
9,10-Di(2-naphthyl)anthracene

CAS Number: 122648-99-1
MDL Number: MFCD00028944
Molecular Formula: C34H22
Molecular Weight: 430.55
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 357

Formula: C₃₄H₂₂
MW: 430.55 g/mol
Melting Pt: 357 °C
MDL Number: MFCD00028944
CAS Number: 122648-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad9c64924b9225bdc75d5eb460f9b8e4ab64d85b&vid=ad9c64924b9225bdc75d5eb460f9b8e4ab64d85b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR