trans-2,6-Difluorocinnamic acid ≥95.0% (by GC)

Supplier: TCI America

D4115-1G D4115-5G
TCD4115-1GEA 60.04 CAD
TCD4115-1G TCD4115-5G
trans-2,6-Difluorocinnamic acid ≥95.0% (by GC)
trans-2,6-Difluorocinnamic acid

CAS Number: 102082-89-3
MDL Number: MFCD00010319
Molecular Formula: C9H6F2O2
Molecular Weight: 184.14
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 122


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: trans-3-(2,6-Difluorophenyl)acrylic Acid

Formula: C₉H₆F₂O₂
Melting Pt: 122 °C
MDL Number: MFCD00010319
CAS Number: 102082-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0119902b5a08b5ffc8e8b6f0eaa21d2037e7f808&vid=0119902b5a08b5ffc8e8b6f0eaa21d2037e7f808&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR