O-Dianisidine dihydrochloride ≥95.0% (by titrimetric analysis) for biochemical research

D3893-5G
TCD3893-1GEA 48.98 CAD
TCD3893-1G TCD3893-5G
O-Dianisidine dihydrochloride ≥95.0% (by titrimetric analysis) for biochemical research
o-Dianisidine dihydrochloride

[for Biochemical Research]
CAS Number: 20325-40-0
MDL Number: MFCD00012488
Molecular Formula: C14H16N2O2
Molecular Weight: 317.21
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 269


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 4,4'-Diamino-3,3'-dimethoxybiphenyl Dihydrochloride, 3,3'-Dimethoxybenzidine Dihydrochloride, Fast Blue B.2HCl

Formula: C₁₄H₁₈Cl₂N₂O₂
Melting Pt: 269 °C
MDL Number: MFCD00012488
CAS Number: 20325-40-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2280f3447c5dfbafa6645892d7cc49a96dc49fda&vid=3bf00fc1f7f18e2a516c3e46b996c4530a281d0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR