2,3-Dimethoxyphenylboronic acid ≥97% (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2,3-Dimethoxybenzeneboronic acid, (2,3-Dimethoxyphenyl)metaboric acid, Dihydroxy(2,3-dimethoxyphenyl)borane

D3853-5G D3853-1G
TCD3853-5GEA 114.08 CAD
TCD3853-5G 76219-694
2,3-Dimethoxyphenylboronic acid ≥97% (contains varying amounts of Anhydride)
2,3-Dimethoxyphenylboronic acid

CAS Number: 40972-86-9
MDL Number: MFCD02683112
Molecular Formula: C8H11BO4
Molecular Weight: 181.98
Form: Crystal
Color: White
Melting point (°C): 68


Other Product Names / Synonyms: 2,3-Dimethoxybenzeneboronic Acid

Formula: C₈H₁₁BO₄
MW: 181.98
Melting Pt: 68 °C
MDL Number: MFCD02683112
CAS Number: 40972-86-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2ed3590bd3a126bee80f7ebb5279e3b3772470f&vid=d2ed3590bd3a126bee80f7ebb5279e3b3772470f&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR