3,4-Dehydro-L-proline ≥99.0% (by HPLC)

Supplier: TCI America

D3825-100MG
TCD3825-100MGEA 218.04 CAD
TCD3825-100MG
3,4-Dehydro-L-proline ≥99.0% (by HPLC)
3,4-Dehydro-L-proline

CAS Number: 4043-88-3
MDL Number: MFCD00065964
Molecular Formula: C5H7NO2
Molecular Weight: 113.12
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 217
Specific rotation [a]20/D: -400 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-3-Pyrroline-2-carboxylic Acid

Formula: C₅H₇NO₂
Melting Pt: 217 °C
MDL Number: MFCD00065964
CAS Number: 4043-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22e156f76d31c9593243e802a510382ce300ac0d&vid=22e156f76d31c9593243e802a510382ce300ac0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR