6,7-Dimethoxy-4-quinazolone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3816-1G D3816-5G
TCD3816-1GEA 92.59 CAD
TCD3816-1G TCD3816-5G
6,7-Dimethoxy-4-quinazolone ≥98.0% (by HPLC, titration analysis)
6,7-Dimethoxy-4-quinazolone

CAS Number: 13794-72-4
MDL Number: MFCD01570147
Molecular Formula: C10H10N2O3
Molecular Weight: 206.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 298


Other Product Names / Synonyms: 6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-3,4-dihydroquinazolin-4-one, 6,7-Dimethoxy-4-hydroxyquinazoline, 6,7-Dimethoxy-4-quinazolinol

Formula: C₁₀H₁₀N₂O₃
Melting Pt: 298 °C
MDL Number: MFCD01570147
CAS Number: 13794-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55f6932a2645d46bc7183d3bade5f0eeac9782a4&vid=55f6932a2645d46bc7183d3bade5f0eeac9782a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR