Dihydrogen di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D3706-100MG
TCD3706-100MGEA 413.96 CAD
TCD3706-100MG
Dihydrogen di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate ≥97.0% (by titrimetric analysis)
Dihydrogen di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate

CAS Number: 386706-31-6
MDL Number: MFCD05662696
Molecular Formula: C32H74Cl2O4P4Pd2
Molecular Weight: 930.58
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 280

Formula: C₃₂H₇₄Cl₂O₄P₄Pd₂
Melting Pt: 280 °C
MDL Number: MFCD05662696
CAS Number: 386706-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb0ac62ce957140d6a8ca6cf6f9a8cd865739102&vid=cb0ac62ce957140d6a8ca6cf6f9a8cd865739102&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR