Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥97.0%

Supplier: TCI America

Synonyms: cis-Dimethyl cyclohex-4-ene-1,2-dicarboxylate, cis-4-Cyclohexene-1,2-dicarboxylic Acid Dimethyl Ester

D3703-5G
TCD3703-5GEA 127.98 CAD
TCD3703-5G
Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥97.0%
Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate

CAS Number: 4841-84-3
MDL Number: MFCD00209581
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 113
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: cis-4-Cyclohexene-1,2-dicarboxylic Acid Dimethyl Ester

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Boiling Pt: 142 °C
Density: 1.15 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00209581
CAS Number: 4841-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba39a459eacafdbac8e1fb973e89b041fe7712a&vid=aba39a459eacafdbac8e1fb973e89b041fe7712a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR