4,5-Difluoro-2-nitroaniline ≥98.0%

Supplier: TCI America
Warning

D3644-25G D3644-5G
TCD3644-25GEA 404.48 CAD
TCD3644-25G TCD3644-5G
4,5-Difluoro-2-nitroaniline ≥98.0%
4,5-Difluoro-2-nitroaniline

CAS Number: 78056-39-0
MDL Number: MFCD00010876
Molecular Formula: C6H4F2N2O2
Molecular Weight: 174.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108

Formula: C₆H₄F₂N₂O₂
Melting Pt: 108 °C
MDL Number: MFCD00010876
CAS Number: 78056-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f4d7980b47161e33c43447995756575926a9a27&vid=9f4d7980b47161e33c43447995756575926a9a27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR