2,5-Dibromobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D3439-5G D3439-25G
TCD3439-5GEA 99.54 CAD
TCD3439-5G TCD3439-25G
2,5-Dibromobenzoic acid ≥97.0% (by GC, titration analysis)
2,5-Dibromobenzoic acid

CAS Number: 610-71-9
MDL Number: MFCD00016494
Molecular Formula: C7H4Br2O2
Molecular Weight: 279.92
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 157

Formula: C₇H₄Br₂O₂
Melting Pt: 157 °C
MDL Number: MFCD00016494
CAS Number: 610-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28af475d1b2554630c88be2d05cca7ccee4c7742&vid=f85d6a8712c29c5a8e5b87953cf50d9be9d226b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR