DL-1,4-Dichloro-2,3-butanediol ≥98.0%

Supplier: TCI America

D3423-5G D3423-25G
TCD3423-5GEA 252.96 CAD
TCD3423-5G TCD3423-25G
DL-1,4-Dichloro-2,3-butanediol ≥98.0%
DL-1,4-Dichloro-2,3-butanediol

CAS Number: 2419-73-0
Molecular Formula: C4H8Cl2O2
Molecular Weight: 159.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 77

Formula: C₄H₈Cl₂O₂
Boiling Pt: 152 °C
Melting Pt: 77 °C
CAS Number: 2419-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37c4ac7258dfda23acafc3c0af7e9fb80aae598e&vid=37c4ac7258dfda23acafc3c0af7e9fb80aae598e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR