2,3-Dimethyl-2,3-butanediamine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

D3212-1G D3212-5G
TCD3212-1GEA 203.82 CAD
TCD3212-1G TCD3212-5G
2,3-Dimethyl-2,3-butanediamine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
2,3-Dimethyl-2,3-butanediamine dihydrochloride

CAS Number: 75804-28-3
Molecular Formula: C6H16N2
Molecular Weight: 189.12
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,3-Diamino-2,3-dimethylbutane Dihydrochloride, 1,1,2,2-Tetramethylethylenediamine Dihydrochloride

Formula: C₆H₁₆N₂
CAS Number: 75804-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce00dd59ee292190caae11241a246bbaec6d0c2b&vid=ce00dd59ee292190caae11241a246bbaec6d0c2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR