N,N-Dibenzylglycine ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: N,N-Dibenzylglycine Ethyl Ester, Di-Bzl-Gly-OEt, Ethyl 2-(dibenzylamino)acetate

D3165-25G D3165-5G
TCD3165-25GEA 636.74 CAD
TCD3165-25G TCD3165-5G
N,N-Dibenzylglycine ethyl ester ≥98.0%
N,N-Dibenzylglycine ethyl ester

CAS Number: 77385-90-1
Molecular Formula: C18H21NO2
Molecular Weight: 283.37
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 56


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Di-Bzl-Gly-OEt

Formula: C₁₈H₂₁NO₂
MW: 283.37 g/mol
Melting Pt: 56 °C
MDL Number: MFCD00796343
CAS Number: 77385-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284aa39b52aa4557bcc0fe7d9ef2fbed85259c0d&vid=284aa39b52aa4557bcc0fe7d9ef2fbed85259c0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR