4,4'-Dichloro-trans-stilbene ≥98.0%

Supplier: TCI America

D3063-1G D3063-250MG
TCD3063-1GEA 733.91 CAD
TCD3063-1G TCD3063-250MG
4,4'-Dichloro-trans-stilbene ≥98.0%
4,4'-Dichloro-trans-stilbene

CAS Number: 1657-56-3
Molecular Formula: C14H10Cl2
Molecular Weight: 249.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 176
Storage Temperature: 0-10°C

Formula: C₁₄H₁₀Cl₂
Boiling Pt: 230 °C
Melting Pt: 176 °C
MDL Number: MFCD01108802
CAS Number: 1657-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e40722283687a73547fd0bfcb5d4831b100a6805&vid=e40722283687a73547fd0bfcb5d4831b100a6805&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR