3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

D2981-1G D2981-5G
TCD2981-1GEA 66.36 CAD
TCD2981-1G TCD2981-5G
3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
3,6-Dibromo-9-phenylcarbazole

CAS Number: 57103-20-5
Molecular Formula: C18H11Br2N
Molecular Weight: 401.10
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal

Formula: C₁₈H₁₁Br₂N
MDL Number: MFCD08276338
CAS Number: 57103-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f730767d965debca7462b5d19c0b148145844bba&vid=f730767d965debca7462b5d19c0b148145844bba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR