Sinapic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

D2932-5G
TCD2932-5GEA 175.22 CAD
TCD2932-5G
Sinapic acid ≥99.0% (by GC, titration analysis)
Sinapic acid

[Matrix for MALDI-TOF/MS]
CAS Number: 530-59-6
MDL Number: MFCD00004401
Molecular Formula: C11H12O5
Molecular Weight: 224.21
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 202


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials, Protein Analysis Reagents
Other Product Names / Synonyms: 4-Hydroxy-3,5-dimethoxycinnamic Acid, Sinapinic Acid

Formula: C₁₁H₁₂O₅
MW: 224.21 g/mol
Boiling Pt: 378
Melting Pt: 202 °C
MDL Number: MFCD00004401
CAS Number: 530-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56dd6b23c766964988ea76e221f994c5d43f7011&vid=aa30dd43f9287a47307819bbb8d70e1bbb16e6f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR