2-(2,4-Dichlorophenyl)ethanol ≥97.0%

Supplier: TCI America

Synonyms: 2,4-Dichlorophenethyl alcohol

D2921-5G
TCD2921-5GEA 79 CAD
TCD2921-5G
2-(2,4-Dichlorophenyl)ethanol ≥97.0%
2-(2,4-Dichlorophenyl)ethanol

CAS Number: 81156-68-5
MDL Number: MFCD00192010
Molecular Formula: C8H8Cl2O
Molecular Weight: 191.05
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110


Other Product Names / Synonyms: 2,4-Dichlorophenethyl Alcohol

Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00192010
CAS Number: 81156-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c9e3cf2ddbd3e2fb3c3ab1d37c9868b7d01d893&vid=8c9e3cf2ddbd3e2fb3c3ab1d37c9868b7d01d893&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR