3,5-Dibromo-2-pyridone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2918-5G
TCD2918-5GEA 102.54 CAD
TCD2918-5G
3,5-Dibromo-2-pyridone ≥98.0% (by GC, titration analysis)
3,5-Dibromo-2-pyridone

CAS Number: 13472-81-6
MDL Number: MFCD00023472
Molecular Formula: C5H3Br2NO
Molecular Weight: 252.89
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210


Other Product Names / Synonyms: 3,5-Dibromo-2-pyridinol, 3,5-Dibromo-2(1H)-pyridinone, 3,5-Dibromo-2-pyridone

Formula: C₅H₃Br₂NO
Melting Pt: 210 °C
MDL Number: MFCD00023472
CAS Number: 13472-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a7ab7d1f98f754dd825ac026242ccad5b37e38&vid=a3a7ab7d1f98f754dd825ac026242ccad5b37e38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR