2,3-Dibromopyridine ≥98.0%

Supplier: TCI America

D2917-1G D2917-5G
TCD2917-1GEA 26.86 CAD
TCD2917-1G TCD2917-5G
2,3-Dibromopyridine ≥98.0%
2,3-Dibromopyridine

CAS Number: 13534-89-9
MDL Number: MFCD00234014
Molecular Formula: C5H3Br2N
Molecular Weight: 236.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 249
Melting point (°C): 59

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 249 °C
Melting Pt: 59 °C
MDL Number: MFCD00234014
CAS Number: 13534-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff5e73b990f8a48b589cf8ab384375c190a1f352&vid=ff5e73b990f8a48b589cf8ab384375c190a1f352&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR