(R)-N,N-Dimethyl-1-[(S)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine ≥98.0% (by GC)

Supplier: TCI America

D2537-100MG
TCD2537-100MGEA 127.35 CAD
TCD2537-100MG
(R)-N,N-Dimethyl-1-[(S)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine ≥98.0% (by GC)
(R)-N,N-Dimethyl-1-[(S)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine

CAS Number: 74311-56-1
MDL Number: MFCD00075284
Molecular Formula: C38H37FeNP2
Molecular Weight: 625.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 141
Specific rotation [a]20/D: -350 deg (C=0.5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R)-(S)-BPPFA

Formula: C₃₈H₃₇FeNP₂
Melting Pt: 141 °C
MDL Number: MFCD00075284
CAS Number: 74311-56-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76059b6f0ae67e161a5ba279bde9a24808bf682a&vid=76059b6f0ae67e161a5ba279bde9a24808bf682a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR