(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine ≥98.0%

Supplier: TCI America

D2520-1G
TCD2520-001GEA 477.16 CAD
TCD2520-001G
(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine ≥98.0%
(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine

CAS Number: 121788-73-6
MDL Number: MFCD01321158
Molecular Formula: C16H14F6N2O4S2
Molecular Weight: 476.41
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 213
Specific rotation [a]20/D: 6 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R,R)-1,2-Bis(trifluoromethanesulfonamido)-1,2-diphenylethane

Formula: C₁₆H₁₄F₆N₂O₄S₂
Melting Pt: 213 °C
MDL Number: MFCD01321158
CAS Number: 121788-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9df6c1c9c3694d6e345f55b5747deedf6d15a207&vid=9df6c1c9c3694d6e345f55b5747deedf6d15a207&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR