Decanenitrile ≥98.0%

Supplier: TCI America

D2426-25ML D2426-500ML D2426-100ML
TCD2426-025MLEA 48.24 CAD
TCD2426-025ML TCD2426-500ML TCD2426-100ML
Decanenitrile ≥98.0%
Decanenitrile

CAS Number: 1975-78-6
MDL Number: MFCD00039505
Molecular Formula: C10H19N
Molecular Weight: 153.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Melting point (°C): -15
Flash Point (°C): 97
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Nonyl Cyanide

Formula: C₁₀H₁₉N
Boiling Pt: 237 °C
Melting Pt: –15 °C
Density: 0.82 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00039505
CAS Number: 1975-78-6
EINECS: 217-830-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad35da6d27d6cab82b2f49c5f80f969e5d53f04d&vid=ad35da6d27d6cab82b2f49c5f80f969e5d53f04d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR