2,3-Dichlorobenzyl alcohol ≥98.0%

Supplier: TCI America
Warning

Synonyms: (2,3-Dichlorophenyl)methanol

D2425-5G
TCD2425-005GEA 101.12 CAD
TCD2425-005G
2,3-Dichlorobenzyl alcohol ≥98.0%
2,3-Dichlorobenzyl alcohol

CAS Number: 38594-42-2
MDL Number: MFCD00238581
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00238581
CAS Number: 38594-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c92ad8abde3aa0abe835af0ec6588a92a9ac712e&vid=c92ad8abde3aa0abe835af0ec6588a92a9ac712e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR