Diphenyl-1-pyrenylphosphine ≥95.0% (by GC)

Supplier: TCI America

D2404-100MG D2404-1G
TCD2404-100MGEA 192.87 CAD
TCD2404-100MG TCD2404-1G
Diphenyl-1-pyrenylphosphine ≥95.0% (by GC)
Diphenyl-1-pyrenylphosphine

CAS Number: 110231-30-6
Molecular Formula: C28H19P
Molecular Weight: 386.43
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 176
Storage Temperature: 0-10°C


Product Categories: Bioscience, Fluorescent Probes, ROS/RNS Detection Reagents, Redox Research Reagents
Other Product Names / Synonyms: 1-(Diphenylphosphino)pyrene, DPPP

Formula: C₂₈H₁₉P
Melting Pt: 176 °C
MDL Number: MFCD00278755
CAS Number: 110231-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4719b6d9ad7d6143caaa66f70ba6620e27af42a&vid=f4719b6d9ad7d6143caaa66f70ba6620e27af42a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR