2,5-Dimethylthiophenol ≥98.0%

Supplier: TCI America
Warning

D2393-5G
TCD2393-005GEA 223.1 CAD
TCD2393-005G
2,5-Dimethylthiophenol ≥98.0%
2,5-Dimethylthiophenol

CAS Number: 4001-61-0
MDL Number: MFCD00010020
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 83
Freezing point (°C): 15
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2,5-Dimethylthiophenol, 2-Mercapto-p-xylene, p-Xylenethiol

Formula: C₈H₁₀S
Boiling Pt: 48…49 °C (0.6 mmHg)
Density: 1.03 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00010020
CAS Number: 4001-61-0
EINECS: 223-649-3
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4664703d73be16598d0065f4209b19dc65aba79d&vid=4664703d73be16598d0065f4209b19dc65aba79d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR