2,6-Dimethylanisole ≥98.0%

Supplier: TCI America

D2383-25ML
TCD2383-25MLEA 85.95 CAD
TCD2383-25ML
2,6-Dimethylanisole ≥98.0%
2,6-Dimethylanisole

CAS Number: 1004-66-6
MDL Number: MFCD00008380
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 67
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 2-Methoxy-m-xylene

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 182
Density: 0.96 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00008380
CAS Number: 1004-66-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdb17540379334ce74a7ac4e63c86646e0bba208&vid=bdb17540379334ce74a7ac4e63c86646e0bba208&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR