Monoethyl dodecanedioate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2374-5G
TCD2374-005GEA 346.18 CAD
TCD2374-005G
Monoethyl dodecanedioate ≥98.0% (by titrimetric analysis)
Monoethyl dodecanedioate

CAS Number: 66003-63-2
MDL Number: MFCD00191643
Molecular Formula: C14H26O4
Molecular Weight: 258.36
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 184
Melting point (°C): 50


Other Product Names / Synonyms: Ethyl Hydrogen Dodecanedioate, Dodecanedioic Acid Monoethyl Ester

Formula: C₁₄H₂₆O₄
Boiling Pt: 184 °C
Melting Pt: 50 °C
MDL Number: MFCD00191643
CAS Number: 66003-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=367a1f7cd43f0c0be5e9d1c77a202a11b3d92ec7&vid=367a1f7cd43f0c0be5e9d1c77a202a11b3d92ec7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR