2,8-Dibromdibenzothiophene ≥96.0%

Supplier: TCI America

D2245-5G D2245-1G D2245-25G
TCD2245-5GEA 175.17 CAD
TCD2245-5G TCD2245-001G TCD2245-25G
2,8-Dibromdibenzothiophene ≥96.0%
2,8-Dibromdibenzothiophene

CAS Number: 31574-87-5
MDL Number: MFCD00092755
Molecular Formula: C12H6Br2S
Molecular Weight: 342.05
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 226

Formula: C₁₂H₆Br₂S
Melting Pt: 226 °C
MDL Number: MFCD00092755
CAS Number: 31574-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73942ccf788427b6cadb2ab6dc1ef7f91c60cd3c&vid=73942ccf788427b6cadb2ab6dc1ef7f91c60cd3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR