Bis(2-pyridylmethyl)amine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2228-5G D2228-25G
TCD2228-005GEA 194.97 CAD
TCD2228-005G TCD2228-025G
Bis(2-pyridylmethyl)amine ≥98.0% (by titrimetric analysis)
Bis(2-pyridylmethyl)amine

CAS Number: 1539-42-0
MDL Number: MFCD00129044
Molecular Formula: C12H13N3
Molecular Weight: 199.26
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 110
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 2,2'-Dipicolylamine, alpha,alpha'-Iminodi(2-picoline)

Formula: C₁₂H₁₃N₃
Boiling Pt: 144 °C
Density: 1.11 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00129044
CAS Number: 1539-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f502810f124bbd69b6c8e929365208188ececde8&vid=f502810f124bbd69b6c8e929365208188ececde8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR