3,5-Difluorobenzonitrile ≥99.0%

Supplier: TCI America

D2081-5G
TCD2081-005GEA 67.78 CAD
TCD2081-005G
3,5-Difluorobenzonitrile ≥99.0%
3,5-Difluorobenzonitrile

CAS Number: 64248-63-1
MDL Number: MFCD00010311
Molecular Formula: C7H3F2N
Molecular Weight: 139.10
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 88


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Cyano-1,3-difluorobenzene

Formula: C₇H₃F₂N
Boiling Pt: 160 °C
Melting Pt: 88 °C
MDL Number: MFCD00010311
CAS Number: 64248-63-1
EINECS: 264-752-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203b3c37f4ef2907cd66d177fd2b95f9a7aab772&vid=203b3c37f4ef2907cd66d177fd2b95f9a7aab772&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR