Dodecanal dimethyl acetal ≥97.0%

Supplier: TCI America

D1937-25ML
TCD1937-025MLEA 164.32 CAD
TCD1937-025ML
Dodecanal dimethyl acetal ≥97.0%
Dodecanal dimethyl acetal

CAS Number: 14620-52-1
MDL Number: MFCD00036645
Molecular Formula: C14H30O2
Molecular Weight: 230.39
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Lauryl Aldehyde Dimethyl Acetal, 1,1-Dimethoxydodecane

Formula: C₁₄H₃₀O₂
Boiling Pt: 133 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00036645
CAS Number: 14620-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2de7c214ccf088d0acb0fcc58baa2e1539a36ab7&vid=2de7c214ccf088d0acb0fcc58baa2e1539a36ab7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR