(2,6-Dichlorophenyl)acetic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2,6-Dichlorophenylacetic acid

D1897-25G
TCD1897-025GEA 211.72 CAD
TCD1897-025G
(2,6-Dichlorophenyl)acetic acid ≥96.0% (by titrimetric analysis)
(2,6-Dichlorophenyl)acetic acid

CAS Number: 6575-24-2
MDL Number: MFCD00004320
Molecular Formula: C8H6Cl2O2
Molecular Weight: 205.03
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 160

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Boiling Pt: 316
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004320
CAS Number: 6575-24-2
EINECS: 229-504-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da195874a351f8738028ce5e0a54596b8712a910&vid=da195874a351f8738028ce5e0a54596b8712a910&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR