N-(2,6-Dimethylphenylcarbamoylmethyl)iminodiacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D1730-1G
TCD1730-001GEA 167.01 CAD
TCD1730-001G
N-(2,6-Dimethylphenylcarbamoylmethyl)iminodiacetic acid ≥98.0% (by HPLC, titration analysis)
N-(2,6-Dimethylphenylcarbamoylmethyl)iminodiacetic acid

CAS Number: 59160-29-1
MDL Number: MFCD00038399
Molecular Formula: C14H18N2O5
Molecular Weight: 294.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Analytical Science, Chelating Agents for Radiopharmaceutical Research
Other Product Names / Synonyms: N-(2,6-Xylidinocarbonylmethyl)iminodiacetic Acid

Formula: C₁₄H₁₈N₂O₅
MDL Number: MFCD00038399
CAS Number: 59160-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b578d00954b3366dc6fc2304c06b4b1c3736d62&vid=4b578d00954b3366dc6fc2304c06b4b1c3736d62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR