2,5-Dichloropyridine ≥99.0%

Supplier: TCI America
Warning

D1607-5G D1607-25G
TCD1607-005GEA 74.26 CAD
TCD1607-005G TCD1607-025G
2,5-Dichloropyridine ≥99.0%
2,5-Dichloropyridine

CAS Number: 16110-09-1
MDL Number: MFCD00006239
Molecular Formula: C5H3Cl2N
Molecular Weight: 147.99
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 98
Melting point (°C): 61
Flash Point (°C): 112

Formula: C₅H₃Cl₂N
Boiling Pt: 98 °C
Melting Pt: 61 °C
Flash Pt: 112 °C
MDL Number: MFCD00006239
CAS Number: 16110-09-1
EINECS: 240-278-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=838a95c03442c796d6fc4b8c9bedeaa35625f568&vid=838a95c03442c796d6fc4b8c9bedeaa35625f568&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR