Dextran, M.W. ~ 40,000

Supplier: TCI America

D1448-500G D1448-25G D1448-100G
TCD1448-500GEA 925.88 CAD
TCD1448-500G TCD1448-025G TCD1448-100G
Dextran, M.W. ~ 40,000
Dextran

CAS Number: 9004-54-0
MDL Number: MFCD00130935
Form: Crystal
Color: White
Specific rotation [a]20/D: 197 deg (C=6, H2O)


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)

Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR