Sodium dodecylbenzenesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

D1428-1G
TCD1428-001GEA 139.66 CAD
TCD1428-001G
Sodium dodecylbenzenesulfonate ≥98.0% (by titrimetric analysis)
Sodium dodecylbenzenesulphonate

CAS Number: 25155-30-0
MDL Number: MFCD00011508
Molecular Formula: C18H30O3S
Molecular Weight: 348.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: linear-Dodecylbenzenesulfonic Acid Sodium Salt, Sodium linear-Dodecylbenzenesulfonate

Formula: C₁₈H₂₉NaO₃S
Melting Pt: > 300 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00011508
CAS Number: 25155-30-0
EINECS: 246-680-4
Merck Index: 13,08686

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR