1,2-Diaminopropane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1329-25G D1329-500G
TCD1329-025GEA 200.98 CAD
TCD1329-025G TCD1329-500G
1,2-Diaminopropane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
1,2-Diaminopropane-N,N,N',N'-tetraacetic acid

CAS Number: 4408-81-5
MDL Number: MFCD00004286
Molecular Formula: C11H18N2O8
Molecular Weight: 306.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 237


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Methyl-EDTA, Propylenediamine-N,N,N',N'-tetraacetic Acid, PDTA

Formula: C₁₁H₁₈N₂O₈
Melting Pt: 237 °C
MDL Number: MFCD00004286
CAS Number: 4408-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08c1195ee0aa6fcc7b09e488c80af231b5277c62&vid=08c1195ee0aa6fcc7b09e488c80af231b5277c62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR