(3,4-Dihydroxyphenyl)acetic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D1283-25G
TCD1283-25GEA 358.66 CAD
TCD1283-25G
(3,4-Dihydroxyphenyl)acetic acid ≥97.0% (by titrimetric analysis)
(3,4-Dihydroxyphenyl)acetic acid

CAS Number: 102-32-9
MDL Number: MFCD00004338
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 4-Carboxymethylcatechol, 4-Carboxymethylpyrocatechol, Catechol-4-acetic Acid, Homoprotocatechuic Acid, Pyrocatechol-4-acetic Acid

Formula: (C₆H₃(OH)₂)CH₂COOH
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 130 °C
MDL Number: MFCD00004338
CAS Number: 102-32-9
EINECS: 203-024-1

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR