ω,ω-Dichloroacetophenone ≥97.0%

Supplier: TCI America

D1122-25G
TCD1122-025GEA 328.64 CAD
TCD1122-025G
ω,ω-Dichloroacetophenone ≥97.0%
ω,ω-Dichloroacetophenone

CAS Number: 2648-61-5
MDL Number: MFCD00000844
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Flash Point (°C): 110
Freezing point (°C): 18
Specific Gravity (20/20): 1.34

Formula: (C₆H₅)C(O)CHCl₂
Boiling Pt: 247 °C
Melting Pt: 23…24 °C
Density: 1.34 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000844
CAS Number: 2648-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50bef846685f7bb140ee0abf079ef7cd882e407a&vid=50bef846685f7bb140ee0abf079ef7cd882e407a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR