Dithiodiglycolic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dithiodiacetic Acid, Dithiodiglycolic Acid

D0945-25G
TCD0945-025GEA 248.06 CAD
TCD0945-025G
Dithiodiglycolic acid ≥96.0% (by titrimetric analysis)
Dithiodiglycolic acid

CAS Number: 505-73-7
MDL Number: MFCD00039607
Molecular Formula: C4H6O4S2
Molecular Weight: 182.21
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 107


Other Product Names / Synonyms: Dithiodiacetic Acid

Formula: C₄H₆O₄S₂
MW: 182.21 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00039607
CAS Number: 505-73-7

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR