Neocuproine hemihydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0771-1G
TCD0771-001GEA 33.18 CAD
TCD0771-001G
Neocuproine hemihydrate ≥98.0% (by HPLC, titration analysis)
Neocuproine hemihydrate

CAS Number: 34302-69-7
MDL Number: MFCD00004973
Molecular Formula: C14H12N2
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 160


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Ligands, Analytical Science, Phenanthrolines (Chelating Agents)
Other Product Names / Synonyms: 2,9-Dimethyl-1,10-phenanthroline

Formula: C₂₈H₂₆N₄O
Melting Pt: 160 °C
MDL Number: MFCD00149306
CAS Number: 34302-69-7
EINECS: 207-601-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3045b104313fec77bca9aeab3c3e9abef5f3cc3&vid=a3045b104313fec77bca9aeab3c3e9abef5f3cc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR