3,3'-Dimethylnaphthidine ≥97% (by titrimetric analysis) for chloride determination

Supplier: TCI America

D0754-5G D0754-1G
TCD0754-5GEA 1304.13 CAD
TCD0754-5G TCD0754-001G
3,3'-Dimethylnaphthidine ≥97% (by titrimetric analysis) for chloride determination
3,3'-Dimethylnaphthidine

[for Colorimetric Determination of Cl in Water]
Form: Crystal
Melting point (°C): 232

Formula: C₂₂H₂₀N₂
Melting Pt: 232 °C
MDL Number: MFCD00042706
CAS Number: 13138-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e7b5097567955e2bb1dcda6727f132d8e9f7d73&vid=192ede38086c2959ce6655c6030a3acaa4bd89f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR