S-[2-(Dimethylamino)ethyl]isothiourea dihydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D0652-10G
TCD0652-010GEA 116.92 CAD
TCD0652-010G
S-[2-(Dimethylamino)ethyl]isothiourea dihydrochloride ≥97.0% (by titrimetric analysis)
S-[2-(Dimethylamino)ethyl]isothiourea dihydrochloride

CAS Number: 16111-27-6
MDL Number: MFCD00060161
Molecular Formula: C5H13N3S
Molecular Weight: 220.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 181

Formula: C₅H₁₃N₃S
Melting Pt: 181 °C
MDL Number: MFCD00060161
CAS Number: 16111-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31c705257242650fab052c7f1a1af95d8a9fb9a0&vid=31c705257242650fab052c7f1a1af95d8a9fb9a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR