5-(4-(Dimethylamino)benzylidene)rhodanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0647-25G D0647-1G
TCD0647-025GEA 772.62 CAD
TCD0647-025G TCD0647-001G
5-(4-(Dimethylamino)benzylidene)rhodanine ≥98.0% (by HPLC, titration analysis)
5-(4-(Dimethylamino)benzylidene)rhodanine

CAS Number: 536-17-4 MDL Number: MFCD00064857 Molecular Formula: C12H12N2OS2 Molecular Weight: 264.36 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 288


Other Product Names / Synonyms: 4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone

Formula: C₁₂H₁₂N₂OS₂
Melting Pt: 288 °C
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3a06ff0af112422350153de5d0506649d835898&vid=2af23c547eaff5252d32800db6bbd26369df19ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR