DL-3,4-Dihydroxymandelic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (±)-3,4-Dihydroxyphenylglycolic acid, (±)-3,4-Dihydroxymandelic acid

D0582-100MG
TCD0582-100MGEA 219.62 CAD
TCD0582-100MG
DL-3,4-Dihydroxymandelic acid ≥98.0% (by titrimetric analysis)
(±)-3,4-Dihydroxymandelic acid

CAS Number: 775-01-9
MDL Number: MFCD00004231
Molecular Formula: C8H8O5
Molecular Weight: 184.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 131
Storage Temperature: 0-10°C

Formula: C₈H₈O₅
MW: 184.15
Melting Pt: 131 °C
MDL Number: MFCD00004231
CAS Number: 14883-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a054d15e6f1c188f6c9b661a3855f669fd60bf99&vid=a054d15e6f1c188f6c9b661a3855f669fd60bf99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR